D-(+)-Galactose ≥99.0%, high purity
Synonyms:
(2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal
, (2R,3S,4S,5R)-Pentahydroxyhexanal
VWRC0637-100GEA
102
SGD
VWRC0637-100G
VWRC0637-500G
D-(+)-Galactose ≥99.0%, high purity
D(+)-Galactose
Formula:
C₆H₁₂O₆ MW: 180,16 g/mol Boiling Pt: 527 °C (1013 hPa) Melting Pt: 163…169 °C Density: 1,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151230 CAS Number: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
Specification Test Results
Purity | ≥99 |
Specific Rotation (2%, Water 5 Hrs.) | 79 – 81° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...