Chicken Native Albumin (from Egg)

ALFAA16951.30EA 0 SGD
ALFAA16951.30
Chicken Native Albumin (from Egg)
CAS No.: 9006-59-1
MDL No.: MFCD00804580
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR