Polyethylene glycol 200
Supplier: Thermo Scientific
ALFAB21918.30EA
55.6
SGD
ALFAB21918.30
ALFAB21918.0B
Polyethylene glycol 200
Polyethylene glycol 200
Formula:
H(OCH₂CH₂)nOH MW: 200 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: –55…–40 °C Density: 1,127 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01779596 CAS Number: 25322-68-3 EINECS: 203-473-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...