Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Phenylbutyric acid ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 90-27-7
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: >110°(230°F)
Refractive Index: 1.5150
Melting Point: 40-44°
Boiling Point: 270-272°
Beilstein Reference: 509876
MDL No.: MFCD00002667

((±)-2-Phenylbutanoic acid)
 
  Formula: C₁₀H₁₂O₂
MW: 164,2 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 39 to 42 °C
CAS Number: 90-27-7
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 100 g ALFAB23222.22
Glass bottle for solids 500 g ALFAB23222.36
Expand All / Collapse All

SPECIFICATIONS

Melting Point 40-44°
Formula Weight 164.20
CAS No. 90-27-7
Purity 98%
Refractive Index 1.5150
Flash Point >110°(230°F)
Boiling Point 270-272°
Molecular Formula C10H12O2
MDL No. MFCD00002667
Beilstein Reference 509876
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty