Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Phenylbutyric acid ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 4593-90-2
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 110°(230°F)
Melting Point: 36-40°
Boiling Point: 159-161°/9mm
Beilstein Reference: 2044322
MDL No.: MFCD00002725

Formula: C₁₀H₁₂O₂
MW: 164,2 g/mol
MDL Number: MFCD00002725
CAS Number: 4593-90-2
EINECS: 224-987-4

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAL08078.14 Unavailable
Supplier Article Number:: L08078.14
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
Expand All / Collapse All


Melting point 36-40°
Formula weight 164.20
CAS No. 4593-90-2
Purity 98%
Flash point >110°(230°F)
Boiling point 159-161°/9mm
Molecular formula C10H12O2
MDL No. MFCD00002725
Beilstein reference 2044322
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty