Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Phenylethyl isothiocyanate ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 4478-92-6
Molecular Formula: C9H9NS
Formula Weight: 163.24
Flash Point: 103°(217°F)
Refractive Index: 1.5810
Odour: Pungent
Boiling Point: 132-133°/20mm
Beilstein Reference: 2207685
UN No.: 2922
MDL No.: MFCD00022061

Formula: C₆H₅CH(NCS)CH₃
MW: 163,24 g/mol
Boiling Pt: 65 °C (0,05 -rr)
Density: 1,06 g/cm³ (20 °C)
MDL Number: MFCD00022061
CAS Number: 4478-92-6

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAB20011.14
Supplier Article Number:: B20011.14
Glass bottle for solids 5 g ALFAB20011.06
Supplier Article Number:: B20011.06
Expand All / Collapse All


Formula weight 163.24
CAS No. 4478-92-6
Purity 97%
Refractive index 1.5810
Flash point 103°(217°F)
Odour/scent Pungent
Boiling point 132-133°/20mm
Molecular formula C9H9NS
MDL No. MFCD00022061
UN No. 2922
Beilstein reference 2207685
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty