Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium (II) chloride hemipentahydrate BAKER ANALYZED® ACS

Supplier: Avantor Performance Materials
Danger



Formula: CdCl₂·2.5H₂O
MW: 228,35 g/mol
Melting Pt: 568 °C
Density: 2,84 g/cm³ (20 °C)
MDL Number: MFCD00799071
CAS Number: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 13,01617
Specification Test Results
Meets ACS Reagent Chemical Requirements
Assay (CdCl₂) (by Ag titrn) 79.5 - 81.0 %
Insoluble Matter <= 0.005 %
Nitrate and Nitrite (as NO₃) <= 0.003 %
Sulfate (SO₄) <= 0.005 %
Ammonium (NH₄) <= 0.002 %
Calcium (Ca) <= 0.005 %
Lead (Pb) <= 0.005 %
Potassium (K) <= 0.02 %
Sodium (Na) <= 0.05 %
Zinc (Zn) <= 0.05 %
Trace Impurities - Copper (Cu) <= 5 ppm
Trace Impurities - Iron (Fe) <= 5 ppm

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle for solids 500 g BAKR1208-01
Supplier Article Number:: 1208-01
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty