Cadmium chloride hydrate ≥99.99% (metals basis)
|
Danger
|
ALFA20129.22EA
509
SGD
ALFA20129.22
ALFA20129.14
Cadmium chloride hydrate ≥99.99% (metals basis)
Cadmium chloride hydrate
Formula:
CdCl₂·H₂O MW: 201,329 g/mol Melting Pt: 565 °C Density: 3,327 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
654054-66-7 EINECS: 233-296-7 UN: 2570 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...