Zinc fluoride tetrahydrate ≥98%
Supplier: Thermo Scientific
|
Danger
|
ALFA14127.36EA
352
SGD
ALFA14127.36
Zinc fluoride tetrahydrate ≥98%
Zinc fluoride tetrahydrate
Formula:
F₂Zn·4H₂O MW: 175,45 g/mol Melting Pt: 872 °C Density: 2,255 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149892 CAS Number: 13986-18-0 EINECS: 232-001-9 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...