Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Histidine

Supplier: Avantor Performance Materials
Synonyms: (S)-(+)-Histidine(S)-2-Amino-3-(4-imidazolyl)propionic acidH-His-OHL-3-Imidazol-4-ylalanineL-HistidineNSC 137773



Formula: C₆H₉N₃O₂
MW: 155,16 g/mol
Melting Pt: 272…273 °C
Density: 1,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://sg.vwr.com/stibo/web/std.lang.all/98/29/2299829.jpg

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 100 g BAKRN327-05
Supplier Article Number:: N327-05
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty