Would you like to log in again?
Biuret Protein Assay Reagent, proteomics grade
Supplier: VWR Chemicals
The Biuret Protein Assay is a simple method for determining protein concentration. The amount of protein present in the sample is calculated based on the colour change of the copper in the Biuret reagent in the presence of peptide bonds. The intensity of the colour change is directly proportional to the number of peptide bonds present.
  • Determine protein concentrations from 1 to 10 mg/ml
  • Independent of amino acid composition
  • Few interfering chemicals
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Biuret Protein Assay Reagent 500 ml VWRCM262-500ML
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty