Pentafluoropropionic acid ≥97%
|
Danger
|
Synonyms:
Perfluoropropionic acid
, Perfluoropropanoic acid
, 2,2,3,3,3-Pentafluoropropionic acid
, Pentafluoropropanoic acid
ALFAA12791.36EA
1870
SGD
ALFAA12791.36
ALFAA12791.14
ALFAA12791.22
Pentafluoropropionic acid ≥97%
Pentafluoropropionic acid
Formula:
C₃HF₅O₂ MW: 164,03 g/mol Boiling Pt: 95…97 °C Density: 1,576 Storage Temperature: Ambient |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 EINECS: 207-021-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...