Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylated spirit (denatured ethanol) 93-97% TECHNICAL

Supplier: VWR Chemicals
Danger

The denaturing agent named Méthylène Régie contains 65 % methyl alcohol + pyrogenic impurities
Delivery information: This product is not available in all countries. Please check with your local VWR I office or supplier.

Formula: H₃CCH₂OH
MW: 46,07 g/mol
Density: 0,81 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
Specification Test Results
Assay (V/V) 93 - 97 %
Colouration Max. 10 APHA
Methylene "Régie" 2 - 6 %

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle 5 l VWRC20827.365 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty