Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexyn-3-ol ≥98%

Supplier: Alfa Aesar

Purity: 98+%
CAS No.: 19780-84-8
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 43°(109°F)
Refractive Index: 1.4430
Boiling Point: 58-59°/25mm
Beilstein Reference: 1698904
UN No.: 1987
MDL No.: MFCD00041593


Formula: C₆H₁₀O
MW: 98,14 g/mol
Boiling Pt: 138 - 139 °C (1013 hPa)
Density: 0,8918 g/cm³ (20 °C)
Storage Temperature: Refrigerator
CAS Number: 19780-84-8
EINECS: 243-304-0
UN: 1987
ADR: 3,III

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAL05437.06
Supplier Article Number:: L05437.06
Glass bottle for solids 1 g ALFAL05437.03
Supplier Article Number:: L05437.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 98.15
CAS No. 19780-84-8
Purity 98+%
Refractive Index 1.4430
Flash point 43°(109°F)
Boiling point 58-59°/25mm
Molecular formula C6H10O
MDL No. MFCD00041593
UN No. 1987
Beilstein reference 1698904
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty