Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-fluorobenzonitrile ≥97%

Supplier: Alfa Aesar
Danger



Formula: C₇H₃ClFN
MW: 155,56 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
CAS Number: 57381-56-3
EINECS: 260-714-5
UN: 3439
ADR: 6.1,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH26817.03
Supplier Article Number:: H26817.03
Glass bottle for solids 5 g ALFAH26817.06
Supplier Article Number:: H26817.06
Glass bottle for solids 25 g ALFAH26817.14
Supplier Article Number:: H26817.14
Expand All / Collapse All

SPECIFICATIONS

Melting point 83-85°
Formula weight 155.56
CAS No. 57381-56-3
Purity 97%
Molecular formula C7H3ClFN
UN No. 3439
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty