Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichloro-5-fluorobenzoyl chloride ≥97%

Supplier: Alfa Aesar
Danger

Purity: 97%
CAS No.: 86393-34-2
Molecular Formula: C7H2Cl3FO
Formula Weight: 227.45
Flash Point: 110°(230°F)
Refractive Index: 1.5730
Boiling Point: 143-144°/35mm
Beilstein Reference: 4989077
UN No.: 3265
MDL No.: MFCD00075341


Formula: C₇H₂Cl₃FO
MW: 227,45 g/mol
Boiling Pt: 143 - 144 °C (35 -rr)
Density: 1,568 g/cm³ (20 °C)
MDL Number: MFCD00075341
CAS Number: 86393-34-2
UN: 3265
ADR: 8,II

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAB23514.06
Supplier Article Number:: B23514.06
Glass bottle for solids 1 g ALFAB23514.03
Supplier Article Number:: B23514.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 227.45
CAS No. 86393-34-2
Purity 97%
Refractive index 1.5730
Flash point >110°(230°F)
Boiling point 143-144°/35mm
Molecular formula C7H2Cl3FO
MDL No. MFCD00075341
UN No. 3265
Beilstein reference 4989077
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty