Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3,6-difluorobenzoyl chloride ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 261762-42-9
Molecular Formula: C7H2Cl2F2O
Formula Weight: 211.00
Refractive Index: 1.5100


Formula: C₇H₂Cl₂F₂O
MW: 210,99 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 70 °C
MDL Number: MFCD01631442
CAS Number: 261762-42-9
UN: 3265
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAL18169.06
Supplier Article Number:: L18169.06
Glass bottle for solids 1 g ALFAL18169.03
Supplier Article Number:: L18169.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 211.00
CAS No. 261762-42-9
Purity 97%
Molecular formula C7H2Cl2F2O
MDL No. MFCD01631442
UN No. 3265
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty