Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-fluorobenzoyl chloride ≥97%

Supplier: Alfa Aesar
Danger



Formula: C₇H₃Cl₂FO
MW: 193 g/mol
Boiling Pt: 82-84 °C/15mm
Storage Temperature: Ambient
MDL Number: MFCD00084954
CAS Number: 21900-54-9
UN: 3265
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAB23439.03
Supplier Article Number:: B23439.03
Glass bottle for solids 5 g ALFAB23439.06
Supplier Article Number:: B23439.06
Glass bottle for solids 25 g ALFAB23439.14
Supplier Article Number:: B23439.14
Expand All / Collapse All

SPECIFICATIONS

Formula weight 193.01
CAS No. 21900-54-9
Purity 97%
Molecular formula C7H3Cl2FO
MDL No. MFCD00084954
UN No. 3265
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping