Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-fluorobenzoyl chloride ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 21900-51-6
Molecular Formula: C7H3Cl2FO
Formula Weight: 193.01
Refractive Index: 1.5500
Boiling Point: 106°/18mm
UN No.: 3265
MDL No.: MFCD01631417

(2-Chloro-5-fluorobenzene-1-carbonyl chloride)
 
  Formula: C₇H₃Cl₂FO
MW: 193 g/mol
Boiling Pt: 106 °C (1013 hPa)
MDL Number: MFCD01631417
CAS Number: 21900-51-6
ADR: 8,II
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAB24369.03
Glass bottle for solids 5 g ALFAB24369.06
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 193.01
CAS No. 21900-51-6
Purity 97%
Refractive Index 1.5500
Boiling Point 106°/18mm
Molecular Formula C7H3Cl2FO
MDL No. MFCD01631417
UN No. 3265
Beilstein Reference 2640754
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty