Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-fluorobenzenesulphonyl chloride ≥97%

Supplier: Alfa Aesar
Synonyms: 3-Chloro-2-fluorobenzene-1-sulphonyl chloride

Purity: 97%
CAS No.: 351003-48-0
Molecular Formula: C6H3Cl2FO2S
Formula Weight: 229.06
Flash Point: 110°(230°F)
Refractive Index: 1.5540
UN No.: 3265
MDL No.: MFCD03094234


Formula: C₆H₃Cl₂FO₂S
MW: 229,06 g/mol
MDL Number: MFCD03094234
CAS Number: 351003-48-0
UN: 3265
ADR: 8,II

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAL19511.06
Supplier Article Number:: L19511.06
Glass bottle for solids 1 g ALFAL19511.03
Supplier Article Number:: L19511.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 229.06
CAS No. 351003-48-0
Purity 97%
Refractive index 1.5540
Flash point >110°(230°F)
Molecular formula C6H3Cl2FO2S
MDL No. MFCD03094234
UN No. 3265
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty