Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-2-fluorobenzenesulphonyl chloride ≥99%

Supplier: Alfa Aesar
Danger
Synonyms: 4-Chloro-2-fluorobenzene-1-sulphonyl chloride



Formula: C₆H₃Cl₂FO₂S
MW: 229,06 g/mol
Melting Pt: 34-38 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094560
CAS Number: 141337-26-0
EINECS: 000-000-0
UN: 3261
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAL19882.06
Supplier Article Number:: L19882.06
Glass bottle for solids 1 g ALFAL19882.03
Supplier Article Number:: L19882.03
Expand All / Collapse All

SPECIFICATIONS

Melting point 34-38°
Formula weight 229.06
CAS No. 141337-26-0
Purity 99%
Flash point >110°(230°F)
Molecular formula C6H3Cl2FO2S
UN No. 3261
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping