Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,4-Trichloro-5-iodobenzene ≥98%

Supplier: Alfa Aesar
Warning



Formula: C₆H₂Cl₃I
MW: 307,34 g/mol
Boiling Pt: 294 °C (110 mmHg)
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00051450
CAS Number: 7145-82-6

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAL06015.06
Supplier Article Number:: L06015.06
Glass bottle for solids 25 g ALFAL06015.14
Supplier Article Number:: L06015.14
Expand All / Collapse All

SPECIFICATIONS

Melting point 102-105°
Formula weight 307.35
CAS No. 7145-82-6
Purity 98%
Molecular formula C6H2Cl3I
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty