2,3-Dichlorophenyl isocyanate ≥97%

Danger

Synonyms: Isocyanic acid 2,3-Dichlorophenyl ester

ALFAL14780.06EA 234 SGD
ALFAL14780.06 ALFAL14780.03
2,3-Dichlorophenyl isocyanate ≥97%
2,3-Dichlorophenyl isocyanate
Formula: C₇H₃Cl₂NO
MW: 188,01 g/mol
Boiling Pt: 58 °C (1 mmHg)
Density: 1,424
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002000
CAS Number: 41195-90-8
EINECS: 255-253-1
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR