Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dichlorophenyl isothiocyanate ≥97%

Supplier: Alfa Aesar
Danger

Purity: 97%
CAS No.: 6590-94-9
Molecular Formula: C7H3Cl2NS
Formula Weight: 204.08
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.6800
Odour: Irritating
Boiling Point: 145-148°/10mm
Beilstein Reference: 511101
UN No.: 2922
MDL No.: MFCD00037828


Formula: C₇H₃Cl₂NS
MW: 204,08 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 60 - 61 °C
Density: 1,422 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00037828
CAS Number: 6590-94-9
EINECS: 229-526-0
UN: 3265
ADR: 8,III

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAA19236.06
Supplier Article Number:: A19236.06
Glass bottle for solids 25 g ALFAA19236.14
Supplier Article Number:: A19236.14
Expand All / Collapse All

SPECIFICATIONS

Formula weight 204.08
CAS No. 6590-94-9
Purity 97%
Refractive index 1.6800
Storage temperature 2°C to 8°C
Flash point >110°(230°F)
Odour/scent Irritating
Boiling point 145-148°/10mm
Molecular formula C7H3Cl2NS
MDL No. MFCD00037828
UN No. 2922
Beilstein reference 511101
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty