3-Nitrobenzenesulphonyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Nitrobenzene-1-sulphonyl chloride
ALFAA17670.22EA
519
SGD
ALFAA17670.22
3-Nitrobenzenesulphonyl chloride ≥98%
3-Nitrobenzenesulphonyl chloride
Formula:
C₆H₄ClNO₄S MW: 221,62 g/mol Melting Pt: 59…63 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007435 CAS Number: 121-51-7 EINECS: 204-476-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...