Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenetole ≥99.5%

Supplier: VWR Chemicals
Warning


Formula: C₆H₅OC₂H₅
MW: 122,17 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: 169,9 °C
Density: 0,956 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
CAS Number: 103-73-1
EINECS: 203-139-7
UN: 3271
ADR: 3,III
Merck Index: 12,07374
Specification Test Results
Assay Min. 99.5 %
Phenol Max. 0.05 %
Water Max. 0.5 %

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle 1 l VWRC27295.294
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty