Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dimethoxythiophenol ≥98%

Supplier: Alfa Aesar
Warning

Purity: 98%
CAS No.: 700-96-9
Molecular Formula: C8H10O2S
Formula Weight: 170.23
Flash Point: 113°(235°F)
Refractive Index: 1.5900
Odour: Stench
Boiling Point: 263°
Beilstein Reference: 1945136
MDL No.: MFCD00052823

Formula: C₈H₁₀O₂S
MW: 170,23 g/mol
Boiling Pt: 86 - 88 °C (0,4 -rr)
Density: 1,19 g/cm³ (20 °C)
MDL Number: MFCD00052823
CAS Number: 700-96-9
UN: 3334
ADR: 9,


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAA14745.06
Supplier Article Number:: A14745.06
Glass bottle for solids 1 g ALFAA14745.03
Supplier Article Number:: A14745.03
Glass bottle for solids 25 g ALFAA14745.14
Supplier Article Number:: A14745.14
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 170.23
CAS No. 700-96-9
Purity 98%
Refractive Index 1.5900
Flash Point >113°(235°F)
Odor/Scent Stench
Boiling Point 263°
Molecular Formula C8H10O2S
MDL No. MFCD00052823
Beilstein Reference 1945136
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty