Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dimethoxybenzenesulphonyl chloride ≥98%

Supplier: Alfa Aesar
Synonyms: 2,5-Dimethoxybenzene-1-sulphonyl chloride

Purity: 98%
CAS No.: 1483-28-9
Molecular Formula: C8H9ClO4S
Formula Weight: 236.68
Melting Point: 111-116°
Beilstein Reference: 2648547
UN No.: 3261
MDL No.: MFCD00024872

Formula: C₈H₉ClO₄S
MW: 236,68 g/mol
Melting Pt: 119 - 121 °C
MDL Number: MFCD00024872
CAS Number: 1483-28-9
UN: 3261

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAA11288.06
Supplier Article Number:: A11288.06
Glass bottle for solids 100 g ALFAA11288.22
Supplier Article Number:: A11288.22
Glass bottle for solids 25 g ALFAA11288.14
Supplier Article Number:: A11288.14
Expand All / Collapse All


Melting point 111-116°
Formula weight 236.68
CAS No. 1483-28-9
Purity 98%
Molecular formula C8H9ClO4S
MDL No. MFCD00024872
UN No. 3261
Beilstein reference 2648547
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty