Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Isopropylthio)benzene ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 3019-20-3
Molecular Formula: C6H5SCH(CH3)2
Formula Weight: 152.26
Refractive Index: 1.5465
Odour: Stench
Boiling Point: 105°/40mm
Beilstein Reference: 1906756
MDL No.: MFCD00039842

Formula: C₉H₁₂S
MW: 152,26 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 56 °C
Density: 0,9859 g/cm³ (20 °C)
CAS Number: 3019-20-3
EINECS: 221-162-0


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 g ALFAA17124.30
Supplier Article Number:: A17124.30
Glass bottle for solids 50 g ALFAA17124.18
Supplier Article Number:: A17124.18
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 152.26
CAS No. 3019-20-3
Purity 98%
Refractive Index 1.5465
Odor/Scent Stench
Boiling point 105°/40mm
Molecular Formula C6H5SCH(CH3)2
MDL No. MFCD00039842
Beilstein reference 1906756
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty