Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Diamino-9,10-anthraquinone ≥90%, Technical Grade

Supplier: Alfa Aesar



Formula: C₁₄H₁₀N₂O₂
MW: 238,25 g/mol
Melting Pt: 260-265 °C
Storage Temperature: Ambient
MDL Number: MFCD00001224
CAS Number: 128-95-0
EINECS: 204-922-6

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAA19156.14
Supplier Article Number:: A19156.14
Glass bottle for solids 100 g ALFAA19156.22
Supplier Article Number:: A19156.22
Glass bottle for solids 500 g ALFAA19156.36
Supplier Article Number:: A19156.36
Expand All / Collapse All

SPECIFICATIONS

Melting point 260-265°
Formula weight 238.25
CAS No. 128-95-0
Purity 90%
Molecular formula C14H10N2O2
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping