Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium dihydrogen phosphate ≥99.5%, Electran Molecular biology grade

Supplier: VWR Chemicals
Synonyms: Potassium dihydrogen orthophosphate



Formula: KH₂PO₄
MW: 136,09 g/mol
Boiling Pt: ~100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744
Specification Test Results
Appearance White crystalline powder
Assay Min. 99.5 %
Heavy metals (as Pb) Max. 0.001 %
Absorbance (260 nm) (0.1 mol/l) Max. 0.05
Absorbance (280 nm) (0,1 mol/l) Max. 0.05
DNases (exo- and endonucleases) Not detected
RNases Not detected
Proteases Not detected

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 g VWRC436053H
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty