Potassium hexachloroplatinate(IV), TECHNICAL
|
Danger
|
Synonyms:
Dipotassium hexachloroplatinate
VWRC26749.081EA
210
SGD
VWRC26749.081
Potassium hexachloroplatinate(IV), TECHNICAL
Potassium hexachloroplatinate(IV)
Formula:
K₂PtCl₆ MW: 485,99 g/mol Melting Pt: 250 °C Density: 3,344 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 EINECS: 240-979-3 UN: 3290 ADR: 6.1 (8),II Merck Index: 12,07800 |
Specification Test Results
Assay (calculated as Pt) | Min. 39.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...