Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium peroxodisulphate ≥99% ACS

Supplier: VWR Chemicals


(Dipotassium peroxodisulphate Peroxydisulphuric acid dipotassium salt Potassium persulphate di-Potassium peroxodisulphate)
R: 8-22-36/37/38-42/43
S: 22-24-26-37
H: H272 H302 H319 H335 H315 H334 H317
P: P210 P280 P285 P302+P352 P304+P340 P305+P351+P338 P309+P311
Danger
 
  Formula: K₂S₂O₈
MW: 270,32 g/mol
Melting Pt: 100 °C
Density: 2,477 g/cm³ (20 °C)
MDL Number: MFCD00011386
CAS Number: 7727-21-1
ADR: 5.1,III
Specification Test Results
Chlorine Compounds < 0.001%
Heavy Metals (as Pb) <= 0.001%
Insolubles <= 0.005%
Iron <= 0.0005%
Manganese <= 0.0002%
Purity >= 99%
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle 500 g VWRC0363-500G
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty