Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum oxide, single crystal

Supplier: Alfa Aesar

CAS No.: 1344-28-1
Molecular Formula: Al2O3
Physical Form: 12.7mm dia x 3.2mm thick, random orientation
MDL No.: MFCD00003424

S: 22
 
  Formula: Al₂O₃
MW: 101,96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
REACH: 01-2119529248-35
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 ALFA40263.KG
5 ALFA40263.KJ
Expand All / Collapse All

SPECIFICATIONS

CAS No. 1344-28-1
Physical Form 12.7mm dia x 3.2mm thick, random orientation
Molecular Formula Al2O3
MDL No. MFCD00003424
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty