Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum oxide substrate, 10x10x1mm, polished one side, R plane

Supplier: Alfa Aesar
Synonyms: Alumina C

Molecular Formula: Al2O3
MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101,96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 ALFA40282.KT
Supplier Article Number:: 40282.KT
5 ALFA40282.KU
Supplier Article Number:: 40282.KU
Expand All / Collapse All


Molecular formula Al2O3
MDL No. MFCD00003424
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty