Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-2-methoxypyridine ≥95%

Supplier: Alfa Aesar

Purity: 95%
CAS No.: 13472-59-8
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Flash Point: 93°(199°F)
Refractive Index: 1.5635
MDL No.: MFCD03095349

R: 22-37/38-41
S: 26-36/37/39
 
  Formula: C₆H₆BrNO
MW: 188,02 g/mol
MDL Number: MFCD03095349
CAS Number: 13472-59-8
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH27279.03
Glass bottle for solids 5 g ALFAH27279.06
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 188.02
CAS No. 13472-59-8
Purity 95%
Refractive Index 1.5635
Flash Point 93°(199°F)
Molecular Formula C6H6BrNO
MDL No. MFCD03095349
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty