Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloronicotinamide ≥98%

Supplier: Alfa Aesar

Purity: 98+%
CAS No.: 10366-35-5
Molecular Formula: C6H5ClN2O
Formula Weight: 156.57
Melting Point: 163-167°
Beilstein Reference: 119026

R: 36/37/38
S: 26-37
H: H319 H335 H315
P: P280 P302+P352 P304+P340 P305+P351+P338 P309+P311
Warning
 
  Formula: C₆H₅ClN₂O
MW: 156,57 g/mol
Melting Pt: 164 to 165 °C
MDL Number: MFCD00006237
CAS Number: 10366-35-5
EINECS: 233-808-9
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAA12090.03
Glass bottle for solids 5 g ALFAA12090.06
Glass bottle for solids 25 g ALFAA12090.14
Expand All / Collapse All

SPECIFICATIONS

Melting Point 163-167°
Formula Weight 156.57
CAS No. 10366-35-5
Purity 98+%
Molecular Formula C6H5ClN2O
MDL No. MFCD00006237
Beilstein Reference 119026
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty