Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloroisonicotinic acid ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 6313-54-8
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: 230-232°
Beilstein Reference: 115861


Formula: C₆H₄ClNO₂
MW: 157,56 g/mol
MDL Number: MFCD00191402
CAS Number: 6313-54-8


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAL19449.03
Supplier Article Number:: L19449.03
Glass bottle for solids 5 g ALFAL19449.06
Supplier Article Number:: L19449.06
Expand All / Collapse All

SPECIFICATIONS

Melting point 230-232°
Formula weight 157.56
CAS No. 6313-54-8
Purity 98%
Molecular formula C6H4ClNO2
MDL No. MFCD00191402
Beilstein reference 115861
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty