Cholesterol (synthetic), Phytochol, GMP

Synonyms: CholesterinCholest-5-en-3-ol(3Beta)-Cholest-5-en-3-ol

RBIP3489-50GEA 13980 SGD
RBIP3489-50G
Cholesterol (synthetic), Phytochol, GMP
Cholesterol
Formula: C₂₇H₄₆O
MW: 386,66 g/mol
MDL Number: MFCD00003646
CAS Number: 57-88-5

Order Now

Specification Test Results

Appearance White crystalline solid
Melting point 147 - 150 °C
Identification TLC Conforms
Color Test Conforms
Identification IR Conforms
Solubility (ethanol) Conforms
Acidity ≤0,3 ml of 0,1 M HCl
Loss on Drying (LOD) ≤0,3%
Sulphated ash ≤0,1%
Assay 98,0 - 102,0%
Purity (area%) ≥99,0%
Specific Rotation < -39.0°
Heavy Metals ≤10 ppm
Residual Solvent Acetone ≤5000 ppm
Residual Solvent Dimethoxyethane ≤100 ppm
Residual Solvent Ethanol ≤5000 ppm
Residual Solvent Ethyl acetate ≤5000 ppm
Residual Solvent Methanol ≤3000 ppm
Residual Solvent Petroleum Ether ≤290 ppm
Residual Solvent Pyridine ≤200 ppm
Residual Solvent Tetrahydrofuran (~ 720 ppm) ≤720 ppm
Bacterial Endotoxin ≤1.0 EU/mg
Total Aerobic Microbial Count ≤100 CFU/g
Total Yeasts and Molds ≤100 CFU/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR