Single-use silicone tubing assemblies
VWRI734-3245EA 498 SGD
VWRI734-3245 VWRI734-3246
Single-use silicone tubing assemblies
Unique features make these tubing assemblies a superior alternative to others.
  • Manufactured in ISO Class 7 cleanroom
  • Supplied gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications.
Assembled in an ISO Class 7 cleanroom and manufactured from USP Class VI materials. Tubing assemblies are offered with custom size and configuration capabilities. All assemblies are individually double-bagged with product label located on inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶.
When used with sterile VWR® Bottle or Flask Assemblies, custom possibilities are endless.