Nickel, 10000 µg/ml (1,00% w/v), BAKER INSTRA-ANALYZED® reagent
BAKR5738-04EA 0 SGD
BAKR5738-04
Nickel, 10000 µg/ml (1,00% w/v), BAKER INSTRA-ANALYZED® reagent
Plasma standard.
  • Prepared from the highest purity raw material 99,999%
  • Assay value accurate within ±0.3% of the assay value
  • Certificate of analysis lists the trace impurity level in µg/ml for over 65 elements