Nickel, 1000 µg/ml (0,10% w/v), single element plasma standard, BAKER INSTRA-ANALYZED® reagent, J.T.Baker®
BAKR5770-04EA 0 SGD
BAKR5770-04
Nickel, 1000 µg/ml (0,10% w/v), single element plasma standard, BAKER INSTRA-ANALYZED® reagent, J.T.Baker®
The standards are meticulously prepared and characterised for ICP and DCP work packaged at concentrations of 1000 µg/ml and 10000 µg/ml in bottles.
  • Prepared from the highest purity raw material 99,999%
  • Assay value accurate within ±0,3% of the assay value
  • Certificate of analysis lists the trace impurity level in µg/ml for over 65 elements