Sterile alcohol for gloves, AquaHol™
Supplier: Micronova
AquaHol™
MICRIC-220EA 490 SGD
MICRIC-220
Sterile alcohol for gloves, AquaHol™
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Custom blend of isopropanol and purified water grade WFI filtered to 0,1 micron.
  • Use on gloves before entering aseptic areas or between critical processes
  • Used in conjunction with the contactless dispenser, it guarantees a constant dose of alcohol
AquaHol™ is provided in double irradiated packaging with certificate in a 500ml bottle. The pump is also irradiated.
The 500ml pumps can be used alone or with the non-contact dispenser.