Flammable solvent spills, SOLUSORB®
BAKR4458-05EA 289 SGD
BAKR4458-05
Flammable solvent spills, SOLUSORB®
Be prepared for solvent spills with SOLUSORB®.
SOLUSORB® provides for safe effective handling of a solvent spill through reduction of flammable or toxic vapors, fast handling, complete cleanup, and convenient disposal.
Ordering information: Available in three sizes or as part of a complete solvent spill clean-up kit (see JT4433-2 series for kits).