Protein-free media, Spodopan
Supplier: PAN-Biotech
PANBP04-850020BEA 7910 SGD
PANBP04-850020B PANBP04-851000 PANBP04-850100 PANBP04-850500 PANBP04-85051S
Protein-free media, Spodopan
Cell Culture Media
Spodopan is a protein-free medium for an optimized growth of insect cells such as Sf9 and Sf21 (Spodoptera frugiperda) in suspension culture. Insect cells are often used for the industrial production of recombinant proteins.
  • Facilitates an easier and more economic purification of final products
  • Guarantees a high cell density with increased production of recombinant proteins (BEVS)
It contains amino acids, vitamins, salts, trace elements, lipids and growth promoting factors in a formulation optimized for insect cells. It contains no protein or any orther components of human or animal origin.
Ideal for for the cultivation of insect cells and the production of recombinant proteins. (e.g.Baculovirus expression vector system, BEVS).
With its protein-free formulation is free of human and animal components. This allows the production of recombinant proteins for medical and therapeutic purposes.