Serum-free media, Panserin 413
Supplier: PAN-Biotech
PANBP04-710413EA 935 SGD
PANBP04-710413 PANBP04-71413S PANBP04-71413
Serum-free media, Panserin 413
Cell Culture Media
Panserin 413 modified is a ready-to-use medium optimised for the serum-free in vitro cultivation of lymphocytes and hybridoma cells.
  • Ready-to-use medium
  • Store at 2 to 8 °C
Based on RPMI 1640/DMEM-F12, trace elements, albumin, cholesterol, soya-lipids and vitamins were added to the medium.
Its modified formulation allows the efficient expansion of T-cells, CIK, LAK and NK cells without the addition of bovine serum or (autologous) human serum.