Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubidium fluoride hydrate ≥99%

Supplier: Alfa Aesar
Danger



Formula: FRb·H₂O
MW: 122,48 g/mol
Boiling Pt: 1410 °C (1013 hPa)
Melting Pt: 775 °C
Density: 3,55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149834
CAS Number: 16422-67-6
EINECS: 236-603-2
UN: 3288
ADR: 6.1,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 10 g ALFAA16846.09
Supplier Article Number:: A16846.09
Glass bottle for solids 50 g ALFAA16846.18
Supplier Article Number:: A16846.18
Expand All / Collapse All

SPECIFICATIONS

Formula weight 104.47(anhy)
CAS No. 16422-67-6
Purity 99%
Odour/scent Odourless
Molecular formula RbF.xH2O
UN No. 3288
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty