Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Biotin

Supplier: Panreac Química SLU

Synonyms: Vitamin B8Vitamin HD-BiotinVitamin B7

PANR203977.1605 PANR203977.1208
D-(+)-Biotin


Formula: C₁₀H₁₆N₂O₃S
MW: 244,31 g/mol
Melting Pt: 232 to 233 °C
Density: 1,41 g/cm³ (20 °C)
Storage Temperature: 2 - 8°C
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
UN: 0000
Merck Index: 13,01231
Order Now


 

ORDER

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty