Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum (III) nitrate hexahydrate

Supplier: Panreac Química SLU
Warning



Formula: La(NO₃)₃·6H₂O
MW: 433,01 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 65 to 67 °C
Density: 2,347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,05363


 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
100 g PANR122669.1208
Supplier Article Number:: 122669.1208
250 g PANR122669.1209
Supplier Article Number:: 122669.1209
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping