Order Entry
Methyl ethyl ketone
  TACTTETH-050
 :  
undefined
Methyl ethyl ketone
  TACTTETH-050
 :  
  1 * 18 L
  • Pk:
    18 L
  • Seq:
    5267
  • Environmentally Preferable:
 : EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
 :
 :72.11 g/mol
 :–4 °C
 :201-159-0
 :1193
 :01-2119457290-43
 :GHS02
 :GHS07
 :Ambient
 :78-93-3
 :C₂H₅COCH₃
 :MFCD00011648
 :79.6
 :–86 °C
 :0.8054 g/cm³ (20 °C)
 :3 II
 :13,06097